Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023560.3 | NP_001018854.2 | 4 | 1092 | ||
NM_001111039.2 | NP_001104509.1 | 4 | 1089 | |||
NM_001287421.1 | NP_001274350.1 | 4 | 578 | |||
NM_001287422.1 | NP_001274351.1 | 2 | 488 | |||
NM_152736.5 | NP_689949.3 | 4 | 687 | |||
XM_011514862.2 | XP_011513164.1 | 4 | 1291 | |||
XM_011514864.1 | XP_011513166.1 | 5 | 868 | |||
XM_011514866.1 | XP_011513168.1 | 4 | 640 | |||
XM_011514867.1 | XP_011513169.1 | 4 | 1004 | |||
XM_011514869.1 | XP_011513171.1 | 5 | 932 | |||
XM_017011264.1 | XP_016866753.1 | 4 | 1289 | |||
XM_017011265.1 | XP_016866754.1 | 4 | 1023 | |||
GenBank mRNA | AK095948.1 | - | 4 | 639 | ||
AK293524.1 | - | 4 | 812 | |||
AK297613.1 | - | 2 | 349 | |||
AK301081.1 | - | 4 | 983 | |||
AK315645.1 | - | 4 | 551 | |||
AL832741.1 | - | 3 | 1280 | |||
BC013951.2 | - | 4 | 543 | 2963683 | ||
BC013962.2 | - | 4 | 543 | 2963683 | ||
BX537535.1 | - | 4 | 960 | |||
DA528922.1 | - | 2 | 349 | |||
DB262147.1 | - | 4 | 554 | |||
Z11773.1 | - | 3 | 609 |