Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277090.1 | NP_001264019.1 | 4 | 830 | ||
NM_001277091.1 | NP_001264020.1 | 4 | 830 | |||
NM_001277092.1 | NP_001264021.1 | 4 | 830 | |||
XM_011526567.2 | XP_011524869.1 | 4 | 827 | |||
XM_011526568.2 | XP_011524870.1 | 4 | 1920 | |||
XM_017026401.1 | XP_016881890.1 | 4 | 827 | |||
GenBank mRNA | AK122867.1 | - | 4 | 831 | ||
AK312802.1 | - | 5 | 782 | |||
AL833214.1 | - | 1 | 399 | |||
BC034810.1 | - | 4 | 806 | 4823435 | ||
BC053858.1 | - | 4 | 804 | 6044705 | ||
BX538032.1 | - | 4 | 817 |