Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEIS2 SIRNA Assays ›
Gene Symbol:
MEIS2
Gene Name:
Meis homeobox 2
Gene Aliases:
HsT18361, MRG1
Chromosome Location:
Chr.15: 36891021 - 37101299 on Build GRCh38
UniGene ID:
Hs.510989
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110298
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEIS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220482.1
NP_001207411.1
13
2659
NM_002399.3
NP_002390.1
13
2610
NM_170674.4
NP_733774.1
13
2888
NM_170675.4
NP_733775.1
12
2813
NM_170676.4
NP_733776.1
12
2792
NM_170677.4
NP_733777.1
13
2909
NM_172315.2
NP_758526.1
12
2385
NM_172316.2
NP_758527.1
13
2675
NR_051953.1
12
2811
XM_006720523.1
XP_006720586.1
12
2203
XM_006720524.1
XP_006720587.1
13
2299
XM_006720525.1
XP_006720588.1
13
2278
XM_006720526.2
XP_006720589.1
12
3199
XM_006720527.2
XP_006720590.1
11
3080
XM_006720529.2
XP_006720592.1
12
3176
XM_011521591.2
XP_011519893.1
10
2265
XM_017022205.1
XP_016877694.1
13
3274
GenBank mRNA
AF017418.1
-
6
1404
AF178948.1
-
13
2489
AF179896.1
-
13
2468
AF179897.1
-
12
2393
AF179898.1
-
12
2372
AF179899.1
-
12
2391
AK001298.1
-
3
1495
AK055936.1
-
12
2109
AK056038.1
-
12
2326
AK226072.1
-
13
2912
BC001516.2
-
13
2614
2964406
BC001844.2
-
13
2614
2964406
BC007202.1
-
13
2614
2964406
BC050431.1
-
13
2539
6066728
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
eye development
transcription from RNA polymerase II promoter
visual learning
response to mechanical stimulus
pancreas development
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
response to growth factor
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transcription corepressor activity
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0