Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173483.3 | NP_775754.2 | 11 | 1408 | ||
XM_011527692.2 | XP_011525994.1 | 12 | 1650 | |||
XM_011527693.2 | XP_011525995.1 | 11 | 1420 | |||
GenBank mRNA | AK096820.1 | - | 11 | 1408 | ||
BC069351.1 | - | 10 | 1345 | 7262372 | ||
BC093894.1 | - | 10 | 1282 | 7939739 | ||
BC093896.1 | - | 10 | 1282 | 7939741 |