Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173582.4 | NP_775853.2 | 8, 9 | 1344 | ||
XM_011544953.2 | XP_011543255.1 | 9, 10 | 1399 | |||
GenBank mRNA | AB019210.1 | - | 8, 9 | 1295 | ||
AK056591.1 | - | 8, 9 | 1302 | |||
BC059360.1 | - | 8, 9 | 1308 | 30332467 |