Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2C2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300945.1 | NP_001287874.1 | 4 | 662 | ||
NM_176880.5 | NP_795361.1 | 4 | 662 | |||
XM_011527688.2 | XP_011525990.1 | 4 | 686 | |||
GenBank mRNA | AK301426.1 | - | 4 | 590 | ||
AY101377.1 | - | 4 | 506 | |||
BC057837.1 | - | 4 | 641 | 30345563 | ||
CF993811.1 | - | 4 | 689 |