Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256584.1 | NP_001243513.1 | 5 | 2005 | ||
NM_002101.4 | NP_002092.1 | 4 | 1187 | |||
NM_016815.3 | NP_058131.1 | 3 | 1130 | |||
XM_006712460.3 | XP_006712523.1 | 4 | 1055 | |||
GenBank mRNA | BC106051.1 | - | 4 | 1019 | 4707035 | |
M28335.1 | - | 4 | 854 | |||
M36284.1 | - | 4 | 860 | |||
X12496.1 | - | 4 | 980 |