Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038493.1 | NP_001033582.1 | 2 | 1424 | ||
NM_178120.4 | NP_835221.2 | 3 | 1624 | |||
GenBank mRNA | BC036189.1 | - | 3 | 1589 | 5311747 | |
BC053351.1 | - | 2 | 1301 | 5243874 |