Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318804.1 | NP_001305733.1 | 4 | 671 | ||
NM_178864.3 | NP_849195.2 | 4 | 671 | |||
XM_017017537.1 | XP_016873026.1 | 6 | 2110 | |||
XM_017017539.1 | XP_016873028.1 | 4 | 671 | |||
GenBank mRNA | AB049469.1 | - | 4 | 596 | ||
AK096253.1 | - | 4 | 671 | |||
BC105001.1 | - | 4 | 584 | 8144004 | ||
BC105003.1 | - | 4 | 584 | 8144006 | ||
BC143630.1 | - | 4 | 584 |