Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM39 SIRNA Assays ›
Gene Symbol:
RBM39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
CAPER, CAPERalpha, FSAP59, HCC1, RNPC2
Chromosome Location:
Chr.20: 35703609 - 35742336 on Build GRCh38
UniGene ID:
Hs.282901
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110538
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM39
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242599.1
NP_001229528.1
16
2032
NM_001242600.1
NP_001229529.1
16
2014
NM_001323422.1
NP_001310351.1
16
2002
NM_001323423.1
NP_001310352.1
19
2243
NM_001323424.1
NP_001310353.1
17
2095
NM_004902.3
NP_004893.1
17
2080
NM_184234.2
NP_909122.1
17
2098
NR_040722.1
18
2171
NR_040723.1
18
2170
NR_040724.1
18
2153
XM_006723890.3
XP_006723953.1
17
3678
XM_006723891.3
XP_006723954.1
18
2701
XM_011529111.2
XP_011527413.1
18
3750
XM_017028138.1
XP_016883627.1
15
1936
XM_017028139.1
XP_016883628.1
19
3067
XM_017028140.1
XP_016883629.1
17
2105
XM_017028141.1
XP_016883630.1
17
3660
XM_017028142.1
XP_016883631.1
18
4556
XM_017028143.1
XP_016883632.1
18
3732
XM_017028144.1
XP_016883633.1
19
2225
XM_017028145.1
XP_016883634.1
17
2087
XM_017028146.1
XP_016883635.1
17
2075
XM_017028147.1
XP_016883636.1
17
1998
GenBank mRNA
AK126158.1
-
14
2257
AK299165.1
-
16
1914
AK299678.1
-
16
1934
AK303910.1
-
17
1970
AL833168.1
-
18
2019
BC107886.1
-
19
1969
BC131543.1
-
16
1798
40085367
BC141835.1
-
17
1948
40148871
BC158172.1
-
17
1948
40148190
BX537770.1
-
18
2090
BX640714.1
-
18
2065
BX640812.1
-
18
2080
CR749443.1
-
17
3765
L10910.1
-
17
1820
L10911.1
-
16
1713
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
mRNA processing
RNA splicing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0