Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000098.2 | NP_000089.1 | 4 | 1039 | ||
XM_005270484.1 | XP_005270541.1 | 4 | 707 | |||
GenBank mRNA | AK223052.1 | - | 2 | 282 | ||
AK312687.1 | - | 4 | 643 | |||
BC002445.2 | - | 4 | 648 | 3346025 | ||
BC005172.2 | - | 4 | 648 | 3605012 | ||
M58581.1 | - | 4 | 625 | |||
U09648.1 | - | 4 | 1040 |