Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000121.3 | NP_000112.1 | 8 | 1604 | ||
NR_033663.1 | 8 | 1854 | ||||
GenBank mRNA | AK315097.1 | - | 8 | 1572 | ||
BC112153.1 | - | 8 | 1546 | 8327621 | ||
BC143697.1 | - | 9 | 1682 | |||
M34986.1 | - | 8 | 1469 | |||
M60459.1 | - | 8 | 1574 |