Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000302.3 | NP_000293.2 | 5 | 654 | ||
NM_001316320.1 | NP_001303249.1 | 6 | 795 | |||
GenBank mRNA | AK294688.1 | - | 5 | 485 | ||
AK299150.1 | - | 6 | 728 | |||
AK312243.1 | - | 5 | 587 | |||
AK316285.1 | - | 6 | 731 | |||
BC016657.2 | - | 5 | 558 | 3917351 | ||
M98252.1 | - | 5 | 565 |