Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000436.3 | NP_000427.1 | 15 | 1740 | ||
XM_017009494.1 | XP_016864983.1 | 14 | 1531 | |||
GenBank mRNA | AK298352.1 | - | 4 | 306 | ||
AK299668.1 | - | 11 | 928 | |||
AK312327.1 | - | 15 | 1436 | |||
AK315902.1 | - | 4 | 388 | |||
BC009001.1 | - | 15 | 1464 | 4148643 | ||
CR457049.1 | - | 15 | 1409 | |||
U62961.1 | - | 15 | 1507 |