Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR1 SIRNA Assays ›
Gene Symbol:
FGFR1
Gene Name:
fibroblast growth factor receptor 1
Gene Aliases:
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Chromosome Location:
Chr.8: 38411138 - 38468834 on Build GRCh38
UniGene ID:
Hs.264887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110745
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174063.1
NP_001167534.1
5, 6
1563
NM_001174064.1
NP_001167535.1
6, 7
1631
NM_001174065.1
NP_001167536.1
5, 6
877
NM_001174066.1
NP_001167537.1
4, 5
616
NM_001174067.1
NP_001167538.1
6, 7
1037
NM_015850.3
NP_056934.2
5, 6
1557
NM_023105.2
NP_075593.1
4, 5
1296
NM_023106.2
NP_075594.1
4, 5
1290
NM_023110.2
NP_075598.2
5, 6
1563
XM_006716303.2
XP_006716366.1
5, 6
1364
XM_006716304.1
XP_006716367.1
5, 6
880
XM_006716306.2
XP_006716369.1
5, 6
1358
XM_006716307.1
XP_006716370.1
5, 6
874
XM_006716309.3
XP_006716372.1
5, 6
1334
XM_006716310.2
XP_006716373.1
4, 5
1097
XM_006716311.1
XP_006716374.1
4, 5
495
XM_006716312.1
XP_006716375.1
4, 5
565
XM_006716313.2
XP_006716376.1
4, 5
1091
XM_006716314.1
XP_006716377.1
4, 5
569
XM_011544443.2
XP_011542745.1
5, 6
869
XM_011544444.1
XP_011542746.1
5, 6
858
XM_011544445.2
XP_011542747.1
5, 6
877
XM_011544446.2
XP_011542748.1
5, 6
864
XM_011544447.2
XP_011542749.1
5, 6
864
XM_011544448.1
XP_011542750.1
4, 5
468
XM_011544449.1
XP_011542751.1
4, 5
462
XM_011544450.2
XP_011542752.1
4, 5
462
XM_011544451.1
XP_011542753.1
3, 4
331
XM_011544452.2
XP_011542754.1
5, 6
952
XM_017013219.1
XP_016868708.1
5, 6
865
XM_017013220.1
XP_016868709.1
5, 6
858
XM_017013221.1
XP_016868710.1
6, 7
1073
XM_017013222.1
XP_016868711.1
5, 6
1364
XM_017013223.1
XP_016868712.1
5, 6
874
XM_017013224.1
XP_016868713.1
5, 6
1358
XM_017013225.1
XP_016868714.1
5, 6
841
XM_017013226.1
XP_016868715.1
4, 5
468
XM_017013227.1
XP_016868716.1
4, 5
462
XM_017013228.1
XP_016868717.1
4, 5
1091
XM_017013230.1
XP_016868719.1
3, 4
422
XM_017013231.1
XP_016868720.1
5, 6
962
GenBank mRNA
AB208919.1
-
6, 7
1418
AK222718.1
-
5, 6
1356
AK223126.1
-
4, 5
1091
AK291754.1
-
5, 6
756
AK292470.1
-
4, 5
616
AK309947.1
-
6, 7
916
BC015035.1
-
6, 7
1291
3911101
BC018128.1
-
5, 6
1321
3896359
BC091494.1
-
3, 4
427
30387634
BX649167.1
-
1, 2
2905
EU826596.1
-
3, 4
348
EU826597.1
-
3, 4
354
FJ809916.1
-
5, 6
1566
FJ809917.1
-
5, 6
1566
KU178066.1
-
4, 5
621
KU178067.1
-
3, 4
354
KU178068.1
-
3, 4
348
KU178069.1
-
3, 4
354
KU178070.1
-
3, 4
348
M34185.1
-
4, 5
609
M34186.1
-
4, 5
874
M34187.1
-
4, 5
770
M34188.1
-
4, 5
870
M34641.1
-
4, 5
624
M37722.1
-
4, 5
576
M60485.1
-
6, 7
1286
M63887.1
-
4, 5
621
M63888.1
-
4, 5
621
M63889.1
-
4, 5
498
X51803.1
-
5, 6
738
X52833.1
-
4, 5
708
X57118.1
-
5, 6
759
X57119.1
-
4, 5
516
X57120.1
-
5, 6
777
X57121.1
-
5, 6
783
X57122.1
-
4, 5
510
X66945.1
-
5, 6
1347
Y00665.1
-
1, 2
21
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-nsq4p:80/10.239.163.53:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0