Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000864.4 | NP_000855.1 | 1 | 1346 | ||
GenBank mRNA | AF498979.1 | - | 1 | 836 | ||
BC007720.2 | - | 1 | 1346 | 4299633 | ||
BT007027.1 | - | 1 | 836 | |||
M81589.1 | - | 1 | 879 |
Set Membership: |
G-protein coupled receptor |