Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251902.1 | NP_001238831.1 | 5 | 744 | ||
NM_003985.4 | NP_003976.2 | 5 | 744 | |||
XM_011524045.2 | XP_011522347.1 | 5 | 792 | |||
XM_011524046.2 | XP_011522348.1 | 5 | 881 | |||
XM_011524047.2 | XP_011522349.1 | 5 | 881 | |||
XM_011524048.1 | XP_011522350.1 | 5 | 649 | |||
GenBank mRNA | AK225403.1 | - | 5 | 627 | ||
AK310684.1 | - | 5 | 627 | |||
BC035782.1 | - | 4 | 609 | 5589312 | ||
U43408.1 | - | 5 | 628 |