Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162407.1 | NP_001155879.1 | 8 | 1378 | ||
NM_004071.3 | NP_004062.2 | 8 | 1098 | |||
NR_027855.1 | 7 | 1007 | ||||
NR_027856.1 | 6 | 2498 | ||||
GenBank mRNA | AK025306.1 | - | 3 | 2405 | ||
AK289365.1 | - | 7 | 959 | |||
AK294295.1 | - | 8 | 1378 | |||
AK316127.1 | - | 8 | 1069 | |||
BC028149.1 | - | 7 | 903 | |||
BC031549.1 | - | 8 | 994 | 4387232 | ||
L29219.1 | - | 8 | 1072 | |||
L29222.1 | - | 7 | 981 | |||
M59287.1 | - | 7 | 982 |