Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130016.2 | NP_001123488.1 | 3 | 415 | ||
NM_001130017.2 | NP_001123489.1 | 3 | 431 | |||
NM_001179.5 | NP_001170.2 | 3 | 415 | |||
XM_005262997.1 | XP_005263054.1 | 2 | 290 | |||
XM_005262999.1 | XP_005263056.1 | 2 | 290 | |||
XM_005263003.1 | XP_005263060.1 | 2 | 290 | |||
XM_005263004.1 | XP_005263061.1 | 2 | 290 | |||
XM_006714220.1 | XP_006714283.1 | 2 | 290 | |||
XM_011531971.1 | XP_011530273.1 | 2 | 290 | |||
XM_017008206.1 | XP_016863695.1 | 2 | 290 | |||
XM_017008207.1 | XP_016863696.1 | 2 | 290 | |||
XM_017008208.1 | XP_016863697.1 | 4 | 803 | |||
XM_017008209.1 | XP_016863698.1 | 2 | 290 | |||
XM_017008210.1 | XP_016863699.1 | 4 | 801 | |||
GenBank mRNA | AK225860.1 | - | 3 | 411 | ||
AK295305.1 | - | 3 | 397 | |||
AK308810.1 | - | 3 | 397 | |||
BC008397.1 | - | 3 | 402 | 4244549 | ||
BC017913.1 | - | 3 | 402 | 4292261 | ||
BT007249.1 | - | 2 | 281 | |||
CR541664.1 | - | 2 | 281 | |||
U47054.1 | - | 3 | 311 |