Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317784.1 | NP_001304713.1 | 16 | 2291 | ||
NM_017988.5 | NP_060458.3 | 16 | 2443 | |||
XM_017019618.1 | XP_016875107.1 | 17 | 2303 | |||
XM_017019619.1 | XP_016875108.1 | 17 | 2455 | |||
XM_017019620.1 | XP_016875109.1 | 18 | 2442 | |||
GenBank mRNA | AB037781.1 | - | 14 | 1609 | ||
AK001597.1 | - | 6 | 603 | |||
AK024274.1 | - | 12 | 1452 | |||
AK027551.1 | - | 12 | 1509 | |||
AK125091.1 | - | 16 | 2428 | |||
AK292980.1 | - | 16 | 2217 | |||
BC012387.2 | - | 7 | 736 | |||
BC063798.1 | - | 16 | 2056 | 6473586 |