Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031847.2 | NP_001027017.1 | 2 | 249 | ||
NM_001876.3 | NP_001867.2 | 2 | 249 | |||
XM_005273762.2 | XP_005273819.1 | 2 | 225 | |||
XM_005273763.1 | XP_005273820.1 | 2 | 225 | |||
XM_017017220.1 | XP_016872709.1 | 2 | 358 | |||
GenBank mRNA | AK172798.1 | - | 2 | 235 | ||
AK309464.1 | - | 2 | 225 | |||
AK314301.1 | - | 2 | 188 | |||
BC000185.1 | - | 2 | 124 | 3352642 | ||
BT009791.1 | - | 1 | 79 | |||
DB063502.1 | - | 2 | 249 | |||
L39211.1 | - | 2 | 155 |