Gene Symbol: ITGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002213.4 | NP_002204.2 | 1, 2 | 383 | ||
XM_005247436.2 | XP_005247493.1 | 1, 2 | 616 | |||
XM_006713630.2 | XP_006713693.1 | 1, 2 | 616 | |||
XM_017006351.1 | XP_016861840.1 | 1, 2 | 154 | |||
XM_017006354.1 | XP_016861843.1 | 1, 2 | 616 | |||
GenBank mRNA | AK315429.1 | - | 1, 2 | 119 | ||
BC006541.2 | - | 1, 2 | 375 | 2958666 | ||
J05633.1 | - | 1, 2 | 124 | |||
M35011.1 | - | 1, 2 | 107 | |||
X53002.1 | - | 1, 2 | 414 |