Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002332.2 | NP_002323.2 | 3 | 702 | ||
XM_017019303.1 | XP_016874792.1 | 3 | 800 | |||
GenBank mRNA | AB209498.1 | - | 2 | 102 | ||
BC021204.2 | - | 3 | 291 | |||
BC045107.1 | - | 3 | 485 | |||
BC052593.1 | - | 3 | 708 | |||
BC072015.1 | - | 3 | 708 | |||
X13916.1 | - | 3 | 702 |