Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204366.1 | NP_001191295.1 | 2 | 390 | ||
NM_001204368.1 | NP_001191297.1 | 2 | 390 | |||
NM_002413.4 | NP_002404.1 | 2 | 390 | |||
XM_017008211.1 | XP_016863700.1 | 2 | 402 | |||
XM_017008212.1 | XP_016863701.1 | 2 | 402 | |||
XM_017008213.1 | XP_016863702.1 | 2 | 402 | |||
GenBank mRNA | AA122237.1 | - | 2 | 390 | ||
AK130948.1 | - | 2 | 339 | |||
BC025416.1 | - | 2 | 304 | 3937750 | ||
BE312976.1 | - | 2 | 354 | |||
CR407640.1 | - | 2 | 138 | |||
DB257575.1 | - | 2 | 154 | |||
U77604.1 | - | 2 | 149 |