Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289155.1 | NP_001276084.1 | 2, 3 | 203 | ||
NM_001289157.1 | NP_001276086.1 | 2, 3 | 203 | |||
NM_002435.2 | NP_002426.1 | 2, 3 | 203 | |||
XM_011521592.1 | XP_011519894.1 | 2, 3 | 279 | |||
XM_011521593.2 | XP_011519895.1 | 2, 3 | 261 | |||
XM_017022207.1 | XP_016877696.1 | 2, 3 | 283 | |||
XM_017022208.1 | XP_016877697.1 | 2, 3 | 261 | |||
GenBank mRNA | AF504648.1 | - | 2, 3 | 168 | ||
AK225149.1 | - | 2, 3 | 166 | |||
AK292374.1 | - | 2, 3 | 156 | |||
AK294030.1 | - | 2, 3 | 172 | |||
BC017351.2 | - | 2, 3 | 152 | 5013257 | ||
BC046357.1 | - | 2, 3 | 158 | 5745956 | ||
BX648475.1 | - | 2, 3 | 180 | |||
CD706167.1 | - | 2, 3 | 194 | |||
X76057.1 | - | 2, 3 | 145 |