Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289155.1 | NP_001276084.1 | 3 | 286 | ||
NM_001289156.1 | NP_001276085.1 | 2 | 158 | |||
NM_001289157.1 | NP_001276086.1 | 3 | 286 | |||
NM_002435.2 | NP_002426.1 | 3 | 286 | |||
XM_011521592.1 | XP_011519894.1 | 3 | 362 | |||
XM_011521593.2 | XP_011519895.1 | 3 | 344 | |||
XM_017022207.1 | XP_016877696.1 | 3 | 366 | |||
XM_017022208.1 | XP_016877697.1 | 3 | 344 | |||
XM_017022209.1 | XP_016877698.1 | 2 | 133 | |||
GenBank mRNA | AF504648.1 | - | 3 | 251 | ||
AK225149.1 | - | 3 | 249 | |||
AK292374.1 | - | 3 | 239 | |||
AK294030.1 | - | 3 | 255 | |||
AK301370.1 | - | 2 | 127 | |||
AK302354.1 | - | 2 | 127 | |||
BC017351.2 | - | 3 | 235 | 5013257 | ||
BC046357.1 | - | 3 | 241 | 5745956 | ||
BX648475.1 | - | 3 | 263 | |||
CD706167.1 | - | 3 | 277 | |||
X76057.1 | - | 3 | 228 |