Gene Symbol: SOAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003101.5 | NP_003092.4 | 2, 3 | 348 | ||
XM_011509911.1 | XP_011508213.1 | 2, 3 | 536 | |||
XM_011509912.1 | XP_011508214.1 | 2, 3 | 343 | |||
GenBank mRNA | AK026611.1 | - | 2, 3 | 260 | ||
BC028940.1 | - | 2, 3 | 180 | 4991365 | ||
DC344271.1 | - | 2, 3 | 348 | |||
L21934.2 | - | 1513 |