Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKACB SIRNA Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
UniGene ID:
Hs.487325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111264
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKACB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242857.2
NP_001229786.1
6
404
NM_001242858.2
NP_001229787.1
3
347
NM_001242859.2
NP_001229788.1
5
395
NM_001242860.2
NP_001229789.1
6
539
NM_001242861.2
NP_001229790.1
5
530
NM_001242862.2
NP_001229791.1
2
143
NM_001300915.1
NP_001287844.1
6
539
NM_001300916.1
NP_001287845.1
3
409
NM_001300917.1
NP_001287846.1
3
347
NM_002731.3
NP_002722.1
3
505
NM_182948.3
NP_891993.1
3
409
NM_207578.2
NP_997461.1
3
505
XM_005271015.3
XP_005271072.1
5
356
XM_005271016.1
XP_005271073.1
5
481
XM_005271017.3
XP_005271074.1
4
346
XM_005271018.4
XP_005271075.1
4
666
XM_005271019.1
XP_005271076.1
3
422
XM_005271020.1
XP_005271077.1
3
389
XM_006710758.1
XP_006710821.1
5
495
XM_017001706.1
XP_016857195.1
6
399
XM_017001707.1
XP_016857196.1
6
399
XM_017001708.1
XP_016857197.1
6
500
XM_017001709.1
XP_016857198.1
5
197
XM_017001710.1
XP_016857199.1
5
495
XM_017001711.1
XP_016857200.1
5
197
XM_017001712.1
XP_016857201.1
5
495
XM_017001713.1
XP_016857202.1
3
447
XM_017001714.1
XP_016857203.1
5
481
XM_017001715.1
XP_016857204.1
5
481
XM_017001716.1
XP_016857205.1
4
349
XM_017001717.1
XP_016857206.1
4
670
XM_017001718.1
XP_016857207.1
3
324
XM_017001719.1
XP_016857208.1
3
422
XM_017001720.1
XP_016857209.1
3
422
GenBank mRNA
AB209189.1
-
6
475
AB451239.1
-
3
154
AB451364.1
-
3
154
AK091420.1
-
5
204
AK296482.1
-
6
206
AK304322.1
-
3
334
AK304375.1
-
3
149
AK314550.1
-
3
401
AK316115.1
-
2
116
BC016285.1
-
3
336
3916100
BC035058.1
-
3
423
5262372
BI598037.1
-
3
219
BM905428.1
-
3
291
BX485407.1
-
3
390
BX537705.1
-
3
386
BX641026.1
-
3
385
BX647515.1
-
6
484
CR936631.1
-
5
198
DA228647.1
-
2
144
DC322547.1
-
6
405
DC418574.1
-
3
409
M34181.1
-
3
201
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0