Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195683.1 | NP_001182612.1 | 5 | 1031 | ||
NM_001195684.1 | NP_001182613.1 | 6 | 867 | |||
NM_003243.4 | NP_003234.2 | 5 | 1031 | |||
NR_036634.1 | 6 | 1128 | ||||
XM_006710867.2 | XP_006710930.1 | 5 | 750 | |||
XM_006710868.2 | XP_006710931.1 | 5 | 716 | |||
GenBank mRNA | AJ251961.1 | - | 4 | 578 | ||
AK225799.1 | - | 5 | 809 | |||
AK291345.1 | - | 5 | 672 | |||
BC099914.1 | - | 8 | 1270 | |||
BC126116.1 | - | 5 | 684 | 8991832 | ||
BC136295.1 | - | 5 | 684 | 9020282 | ||
BC143869.1 | - | 6 | 781 | |||
HG764816.1 | - | 6 | 867 | |||
L07594.1 | - | 6 | 861 |