Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008566.1 | NP_001008566.1 | 3 | 823 | ||
NM_003595.3 | NP_003586.3 | 3 | 715 | |||
XM_006724338.3 | XP_006724401.1 | 4 | 834 | |||
GenBank mRNA | AF049891.1 | - | 3 | 679 | ||
AF061254.1 | - | 1 | 487 | |||
AJ006198.1 | - | 1 | 569 | |||
AK074538.1 | - | 3 | 715 | |||
AK075139.1 | - | 3 | 787 | |||
AK304545.1 | - | 5 | 799 | |||
AL136623.2 | - | 3 | 699 | |||
BC001057.2 | - | 3 | 694 | 3509627 | ||
BC017509.1 | - | 3 | 679 | 4857366 | ||
CR456597.1 | - | 1 | 577 | |||
CR533528.1 | - | 1 | 482 |