Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008566.1 | NP_001008566.1 | 7 | 1649 | ||
NM_003595.3 | NP_003586.3 | 7 | 1541 | |||
XM_006724338.3 | XP_006724401.1 | 8 | 1660 | |||
GenBank mRNA | AF049891.1 | - | 7 | 1505 | ||
AJ006198.1 | - | 5 | 1394 | |||
AK057171.1 | - | 1 | 2168 | |||
AK074538.1 | - | 7 | 1541 | |||
AK075139.1 | - | 7 | 1613 | |||
AK304545.1 | - | 9 | 1625 | |||
AL136623.2 | - | 7 | 1525 | |||
BC001057.2 | - | 7 | 1520 | 3509627 | ||
BC017509.1 | - | 7 | 1505 | 4857366 |