Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003842.4 | NP_003833.4 | 2 | 452 | ||
NM_147187.2 | NP_671716.2 | 2 | 452 | |||
GenBank mRNA | AB451272.1 | - | 2 | 159 | ||
AB451399.1 | - | 2 | 159 | |||
AF012535.1 | - | 2 | 298 | |||
AF012628.1 | - | 2 | 159 | |||
AF016266.1 | - | 2 | 276 | |||
AF016268.1 | - | 2 | 268 | |||
AF016849.1 | - | 2 | 303 | |||
AF018657.1 | - | 2 | 159 | |||
AF018658.1 | - | 2 | 159 | |||
AF020501.1 | - | 2 | 159 | |||
AF022386.1 | - | 2 | 159 | |||
AF153687.1 | - | 2 | 277 | |||
AF192548.1 | - | 2 | 259 | |||
AK291216.1 | - | 2 | 279 | |||
AL529460.3 | - | 2 | 452 | |||
AY358277.1 | - | 2 | 298 | |||
BC001281.1 | - | 2 | 306 | 3458466 | ||
BX538104.1 | - | 2 | 292 | |||
CR541898.1 | - | 2 | 159 | |||
DA738472.1 | - | 2 | 452 | |||
DA889488.1 | - | 2 | 296 |