Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KYAT1 SIRNA Assays ›
Gene Symbol:
KYAT1
Gene Name:
kynurenine aminotransferase 1
Gene Aliases:
CCBL1, GTK, KAT1, KATI
Chromosome Location:
Chr.9: 128832942 - 128882551 on Build GRCh38
UniGene ID:
Hs.495250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111419
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KYAT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122671.1
NP_001116143.1
12
1360
NM_001122672.1
NP_001116144.1
11
1216
NM_001287390.1
NP_001274319.1
14
1757
NM_004059.4
NP_004050.3
12
1366
NR_109829.1
12
1398
XM_006717311.3
XP_006717374.1
13
1552
XM_011519165.2
XP_011517467.1
14
1728
XM_011519167.2
XP_011517469.2
12
1332
XM_011519168.1
XP_011517470.1
11
1191
XM_011519169.1
XP_011517471.1
12
1460
XM_011519170.2
XP_011517472.1
12
1828
XM_017015260.1
XP_016870749.1
13
1512
XM_017015261.1
XP_016870750.1
14
1734
XM_017015262.1
XP_016870751.1
14
2223
XM_017015263.1
XP_016870752.1
14
1638
XM_017015264.1
XP_016870753.1
13
1545
XM_017015265.1
XP_016870754.1
12
1922
XM_017015266.1
XP_016870755.1
13
1553
XM_017015267.1
XP_016870756.1
12
1374
XM_017015268.1
XP_016870757.1
13
1536
GenBank mRNA
AK094505.1
-
12
1941
AK291178.1
-
12
1354
AK297995.1
-
14
1613
AK314427.1
-
12
1236
BC021262.2
-
12
1359
4137942
BC033685.1
-
11
1070
5193675
X82224.1
-
11
1181
Back To Top
More Information
Panther Classification:
Molecular Function -
transaminase
Gene Ontology Categories:
Function(s)
Process(es)
L-phenylalanine catabolic process
tryptophan catabolic process
cellular modified amino acid metabolic process
cellular amino acid biosynthetic process
kynurenine metabolic process
L-kynurenine catabolic process
protein binding
transaminase activity
kynurenine-oxoglutarate transaminase activity
pyridoxal phosphate binding
protein homodimerization activity
L-phenylalanine:pyruvate aminotransferase activity
glutamine-phenylpyruvate transaminase activity
cysteine-S-conjugate beta-lyase activity
L-glutamine:pyruvate aminotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0