Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASMTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173473.1 | NP_001166944.1 | 6 | 479 | ||
NM_001173474.1 | NP_001166945.1 | 5 | 516 | |||
NM_004192.3 | NP_004183.2 | 6 | 564 | |||
XM_005274434.3 | XP_005274491.1 | 7 | 670 | |||
XM_011546185.2 | XP_011544487.1 | 4 | 312 | |||
XM_017029919.1 | XP_016885408.1 | 5 | 360 | |||
GenBank mRNA | AK090498.1 | - | 5 | 468 | ||
AK225770.1 | - | 6 | 486 | |||
AK297805.1 | - | 5 | 397 | |||
AK299231.1 | - | 6 | 482 | |||
AK301844.1 | - | 6 | 473 | |||
BC002508.2 | - | 6 | 450 | 3051194 | ||
BC010089.2 | - | 6 | 448 | 2988425 | ||
BM129217.1 | - | 1 | 13 | |||
DA301876.1 | - | 6 | 519 | |||
DA904467.1 | - | 6 | 481 |