Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004491.4 | NP_004482.4 | 1 | 293 | ||
XM_011526873.2 | XP_011525175.1 | 2 | 689 | |||
XM_017026714.1 | XP_016882203.1 | 2 | 689 | |||
GenBank mRNA | AB051509.2 | - | 1 | 293 | ||
AF159851.1 | - | 1 | 293 | |||
BC131565.1 | - | 1 | 293 | |||
BC150257.1 | - | 1 | 301 | 9007187 |