Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004508.3 | NP_004499.2 | 1, 2 | 502 | ||
GenBank mRNA | AK222875.1 | - | 1, 2 | 22 | ||
AK303669.1 | - | 1, 2 | 256 | |||
BC107893.1 | - | 1, 2 | 26 | |||
BE891119.1 | - | 1, 2 | 306 | |||
X17025.1 | - | 1, 2 | 21 |