Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SHMT2 SIRNA Assays ›
Gene Symbol:
SHMT2
Gene Name:
serine hydroxymethyltransferase 2
Gene Aliases:
GLYA, HEL-S-51e, SHMT
Chromosome Location:
Chr.12: 57229573 - 57234935 on Build GRCh38
UniGene ID:
Hs.741179
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111557
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SHMT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166356.1
NP_001159828.1
11
1472
NM_001166357.1
NP_001159829.1
11
1564
NM_001166358.1
NP_001159830.1
11
1463
NM_001166359.1
NP_001159831.1
11
1356
NM_005412.5
NP_005403.2
11
1502
NR_029415.1
10
1304
NR_029416.1
10
1383
NR_029417.1
10
1422
NR_048562.1
10
1512
XM_011538675.2
XP_011536977.1
11
1442
XM_011538676.2
XP_011536978.1
11
1433
XM_011538677.2
XP_011536979.1
10
1343
XM_011538678.2
XP_011536980.1
10
1325
GenBank mRNA
AK055053.1
-
6
2705
AK223555.1
-
11
1364
AK295941.1
-
10
1358
AK296183.1
-
11
1316
AK297173.1
-
10
1264
AK298231.1
-
10
1157
AK301339.1
-
11
1142
AK301443.1
-
10
1326
AK303827.1
-
10
1178
AK315916.1
-
11
1556
BC011911.2
-
11
1306
4139001
BC013677.2
-
11
1366
3863368
BC025355.1
-
8
889
BC032584.1
-
11
1345
5499481
BC044211.1
-
11
1342
5521082
BC091501.1
-
10
1226
BT006866.1
-
11
1296
BX647711.1
-
11
1423
BX648847.1
-
9
904
EU668344.1
-
11
1306
KU178333.1
-
11
1296
KU178334.1
-
10
932
L11932.1
-
10
1207
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
glycine metabolic process
L-serine metabolic process
L-serine biosynthetic process
one-carbon metabolic process
positive regulation of cell proliferation
glycine biosynthetic process from serine
response to type I interferon
tetrahydrofolate interconversion
tetrahydrofolate metabolic process
folic acid metabolic process
protein tetramerization
protein homotetramerization
protein K63-linked deubiquitination
chromatin binding
glycine hydroxymethyltransferase activity
protein binding
L-allo-threonine aldolase activity
amino acid binding
pyridoxal phosphate binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0