Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002152.2 | NP_002143.1 | 1 | 465 | ||
XM_017026733.1 | XP_016882222.1 | 1 | 484 | |||
GenBank mRNA | BC069795.1 | - | 1 | 448 | 7262494 | |
BC069802.1 | - | 1 | 448 | 7262506 | ||
BC094691.1 | - | 1 | 465 | 30529366 | ||
BC112355.1 | - | 1 | 430 | 40034092 | ||
M60052.1 | - | 1 | 448 |