Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006941.2 | NP_001006942.1 | 3, 4 | 556 | ||
NM_005787.5 | NP_005778.1 | 3, 4 | 483 | |||
NR_024533.1 | 2, 3 | 383 | ||||
NR_024534.1 | 3, 4 | 446 | ||||
XM_011512322.1 | XP_011510624.1 | 3, 4 | 368 | |||
XM_011512323.2 | XP_011510625.1 | 3, 4 | 354 | |||
GenBank mRNA | AK289361.1 | - | 3, 4 | 512 | ||
AK297701.1 | - | 2, 3 | 337 | |||
AK299575.1 | - | 2, 3 | 374 | |||
BC002839.2 | - | 3, 4 | 458 | 3636604 | ||
BC004313.1 | - | 3, 4 | 438 | |||
BC021062.1 | - | 3, 4 | 447 | |||
BM920198.1 | - | 3, 4 | 412 | |||
BX339649.2 | - | 3, 4 | 561 | |||
KP189279.1 | - | 3, 4 | 476 | |||
KP189280.1 | - | 3, 4 | 439 | |||
KP189281.1 | - | 2, 3 | 292 | |||
KP189282.1 | - | 3, 4 | 593 | |||
KP189283.1 | - | 2, 3 | 376 | |||
KP189284.1 | - | 3, 4 | 474 | |||
KP189285.1 | - | 3, 4 | 437 | |||
KP189286.1 | - | 2, 3 | 292 | |||
KP189287.1 | - | 3, 4 | 392 | |||
KP189288.1 | - | 2, 3 | 374 | |||
KP189289.1 | - | 3, 4 | 473 | |||
KP189290.1 | - | 3, 4 | 437 | |||
KP189291.1 | - | 2, 3 | 292 | |||
KP189292.1 | - | 3, 4 | 392 | |||
KP189293.1 | - | 2, 3 | 374 | |||
KP189294.1 | - | 3, 4 | 452 | |||
KP189295.1 | - | 3, 4 | 452 | |||
KP189296.1 | - | 3, 4 | 415 | |||
KP189297.1 | - | 3, 4 | 415 | |||
KP189299.1 | - | 3, 4 | 308 | |||
KP189300.1 | - | 2, 3 | 352 | |||
KP189301.1 | - | 3, 4 | 415 | |||
KP189302.1 | - | 3, 4 | 473 | |||
KP189303.1 | - | 3, 4 | 435 | |||
KP189304.1 | - | 2, 3 | 292 | |||
KP189305.1 | - | 3, 4 | 392 | |||
KP189306.1 | - | 2, 3 | 372 | |||
KP189307.1 | - | 3, 4 | 472 | |||
KP189308.1 | - | 3, 4 | 435 | |||
KP189309.1 | - | 2, 3 | 292 | |||
KP189310.1 | - | 3, 4 | 392 | |||
KP189311.1 | - | 2, 3 | 372 | |||
KP189312.1 | - | 3, 4 | 472 | |||
KP189313.1 | - | 3, 4 | 472 | |||
KP189314.1 | - | 3, 4 | 472 | |||
KP189315.1 | - | 3, 4 | 435 | |||
KP189316.1 | - | 3, 4 | 435 | |||
KP189317.1 | - | 3, 4 | 435 | |||
KP189318.1 | - | 2, 3 | 292 | |||
KP189319.1 | - | 2, 3 | 292 | |||
KP189320.1 | - | 2, 3 | 292 | |||
KP189321.1 | - | 3, 4 | 392 | |||
KP189322.1 | - | 3, 4 | 392 | |||
KP189324.1 | - | 2, 3 | 372 | |||
KP189325.1 | - | 2, 3 | 372 | |||
KP189326.1 | - | 2, 3 | 372 | |||
Y09022.1 | - | 3, 4 | 483 |