Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006456.2 | NP_006447.2 | 9 | 1528 | ||
XM_005256954.3 | XP_005257011.1 | 8 | 1447 | |||
GenBank mRNA | AJ251053.1 | - | 9 | 1312 | ||
AK225943.1 | - | 9 | 1326 | |||
BC040455.1 | - | 9 | 1310 | 4252792 | ||
U14550.1 | - | 9 | 1312 |