Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278318.1 | NP_001265247.1 | 8 | 1281 | ||
NM_006863.3 | NP_006854.1 | 10 | 1881 | |||
NR_103501.1 | 9 | 1683 | ||||
GenBank mRNA | AF025529.1 | - | 8 | 1197 | ||
AF025530.1 | - | 10 | 1809 | |||
AK290919.1 | - | 10 | 1825 |