Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AK1 SIRNA Assays ›
Gene Symbol:
AK1
ST6GALNAC6
Gene Name:
adenylate kinase 1
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
Gene Aliases:
HTL-S-58j
SIAT7-F, SIAT7F, ST6GALNACVI
Chromosome Location:
Chr.9: 127866480 - 127886749 on Build GRCh38
Chromosome Location:
Chr.9: 127885321 - 127906725 on Build GRCh38
UniGene ID:
Hs.175473
Hs.109672
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
111846
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017014427.1
XP_016869916.1
1
290
Gene Symbol: ST6GALNAC6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286999.1
NP_001273928.1
7
1312
NM_001287000.1
NP_001273929.1
6
1261
NM_001287001.1
NP_001273930.1
6
1286
NM_001287002.1
NP_001273931.1
6
1257
NM_001287003.1
NP_001273932.1
5
879
NM_013443.4
NP_038471.2
7
1316
NR_104629.1
6
1234
XM_011518610.2
XP_011516912.1
8
2057
XM_011518611.2
XP_011516913.1
7
2002
XM_017014658.1
XP_016870147.1
6
1902
XM_017014659.1
XP_016870148.1
8
2053
XM_017014660.1
XP_016870149.1
6
1906
XM_017014661.1
XP_016870150.1
7
1998
XM_017014662.1
XP_016870151.1
6
1232
XM_017014663.1
XP_016870152.1
7
1392
XM_017014664.1
XP_016870153.1
6
1282
XM_017014665.1
XP_016870154.1
7
1323
XM_017014666.1
XP_016870155.1
7
1396
XM_017014667.1
XP_016870156.1
6
1667
GenBank mRNA
AK023900.1
-
6
1212
AK025959.1
-
3
655
AK057100.1
-
6
1286
AK226174.1
-
6
1142
AK300534.1
-
5
761
AY358672.1
-
6
1222
BC006564.2
-
7
1233
3009935
BC007802.2
-
7
1260
4128833
BC016299.2
-
7
1260
3507812
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
nucleoside diphosphate phosphorylation
ADP biosynthetic process
cell cycle arrest
nucleoside triphosphate biosynthetic process
nucleobase-containing small molecule interconversion
AMP metabolic process
ATP metabolic process
ganglioside biosynthetic process
glycosylceramide metabolic process
glycosphingolipid metabolic process
glycoprotein metabolic process
oligosaccharide metabolic process
oligosaccharide biosynthetic process
cell-cell recognition
protein N-linked glycosylation via asparagine
sialylation
adenylate kinase activity
nucleoside diphosphate kinase activity
ATP binding
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0