Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002209.2 | NP_002200.2 | 5, 6 | 628 | ||
XM_005255313.1 | XP_005255370.1 | 5, 6 | 548 | |||
XM_011545849.1 | XP_011544151.1 | 5, 6 | 561 | |||
GenBank mRNA | AK314246.1 | - | 5, 6 | 582 | ||
Y00796.1 | - | 5, 6 | 540 |