Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018361.3 | NP_060831.2 | 5 | 847 | ||
GenBank mRNA | AF375789.1 | - | 5 | 809 | ||
AK002072.1 | - | 5 | 511 | |||
AL136587.1 | - | 5 | 537 | |||
BC023550.2 | - | 5 | 791 | 4129973 | ||
BC068519.1 | - | 5 | 511 | |||
BC080537.1 | - | 5 | 816 | 5015128 |