Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289107.1 | NP_001276036.1 | 10 | 2364 | ||
NM_018414.4 | NP_060884.1 | 9 | 2227 | |||
NR_110309.1 | 8 | 2123 | ||||
GenBank mRNA | AK000113.1 | - | 8 | 1381 | ||
AY358918.1 | - | 9 | 2172 | |||
BC022462.1 | - | 8 | 1978 | 4816713 | ||
Y11339.2 | - | 9 | 2087 |