Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243794.1 | NP_001230723.1 | 2 | 1419 | ||
NM_001243795.1 | NP_001230724.1 | 2 | 1433 | |||
NM_018641.4 | NP_061111.1 | 2 | 1447 | |||
XM_011515443.2 | XP_011513745.1 | 2 | 1572 | |||
XM_011515444.2 | XP_011513746.1 | 2 | 1395 | |||
GenBank mRNA | AF239822.1 | - | 2 | 1358 | ||
AJ289131.1 | - | 1 | 1345 | |||
AK223191.1 | - | 2 | 1368 | |||
AK223211.1 | - | 2 | 1368 | |||
AY358574.1 | - | 2 | 1383 | |||
BC002918.1 | - | 2 | 1400 | 3955396 | ||
BC015954.1 | - | 2 | 1400 | |||
BC095492.1 | - | 2 | 1407 | 30511863 |