Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244949.1 | NP_001231878.1 | 11 | 1306 | ||
NM_020918.5 | NP_065969.3 | 11 | 1302 | |||
XM_005269998.1 | XP_005270055.1 | 11 | 1447 | |||
GenBank mRNA | AB046780.1 | - | 5 | 594 | ||
AK172799.1 | - | 9 | 1135 | |||
AL832464.1 | - | 11 | 1258 | |||
AL833061.2 | - | 11 | 1288 | |||
AL833093.2 | - | 11 | 1291 | |||
BC030783.1 | - | 11 | 1270 | 4821342 | ||
BX647451.1 | - | 11 | 1270 |