Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022166.3 | NP_071449.1 | 5, 6 | 1382 | ||
XM_017023539.1 | XP_016879028.1 | 5, 6 | 1435 | |||
XM_017023540.1 | XP_016879029.1 | 5, 6 | 1435 | |||
GenBank mRNA | AJ277441.1 | - | 3, 4 | 901 | ||
AJ539163.1 | - | 5, 6 | 1382 |