Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MOGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025098.3 | NP_079374.2 | 3, 4 | 553 | ||
XM_011545267.1 | XP_011543569.1 | 3, 4 | 553 | |||
GenBank mRNA | AK026297.1 | - | 3, 4 | 552 | ||
AK128620.1 | - | 2, 3 | 1010 | |||
AK291998.1 | - | 3, 4 | 529 | |||
AY157608.1 | - | 3, 4 | 483 | |||
BC103876.1 | - | 3, 4 | 483 | 40004273 | ||
BC103877.1 | - | 3, 4 | 483 | 40004282 | ||
BC103878.1 | - | 3, 4 | 483 | 40004283 |