Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002279.4 | NP_002270.1 | 7 | 1401 | ||
GenBank mRNA | BC009971.2 | - | 7 | 1401 | 4310076 | |
X82634.1 | - | 7 | 1223 |